missing translation for 'onlineSavingsMsg'
Learn More

Capmatinib, MedChemExpress

Product Code. 16478357
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478357 5 mg 5mg
16491827 50 mg 50mg
16471076 200 mg 200mg
16466176 100 mg 100mg
16425668 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16478357 Supplier Medchem Express Supplier No. HY13404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Capmatinib (INC280; INCB28060) is a potent, orally active, selective, and ATP competitive c-Met kinase inhibitor (IC50=0.13 nM).

Capmatinib can inhibit phosphorylation of c-MET as well as c-MET pathway downstream effectors such as ERK1/2, AKT, FAK, GAB1, and STAT3/5. Capmatinib potently inhibits c-MET-dependent tumor cell proliferation and migration and effectively induces apoptosis. Antitumor activity. Capmatinib is largely metabolized by CYP3A4 and aldehyde oxidase.

Specifications

Chemical Name or Material Capmatinib
CAS 1029712-80-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H17FN6O
Quantity 5 mg
Synonym INC280 INCB28060
Solubility Information DMSO : 25 mg/mL (60.62 mM; Need ultrasonic) ∣H2O : 10 mg/mL (24.25 mM; Need ultrasonic)
SMILES O=C(NC)C1=CC=C(C2=NN3C(N=C2)=NC=C3CC4=CC=C5N=CC=CC5=C4)C=C1F
Molecular Weight (g/mol) 412.42
Formula Weight 412.42
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.