missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 0.5M buffer soln., pH 10.5

Product Code. 15494819
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15494819

Brand: Thermo Scientific Chemicals J62446.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

CAPS (N-cyclohexyl-3-aminopropanesulfonic acid) buffer salt is used to formulate CAPS buffer, a zwitterionic buffer that is useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electrotransfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI > 8.5. CAPS is not one of the original Good’s buffers, although it has a similar structure to the other propanesulfonic acids and was selected as a highly water soluble buffering reagent with an optimum buffering pH of 10.4 and minimal reactivity with enzymes or proteins, minimal salt effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CAPS (N-cyclohexyl-3-aminopropanesulfonic acid) buffer salt is used to formulate CAPS buffer, a zwitterionic buffer that is useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electrotransfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI>8.5. CAPS is not one of the original Good’s buffers, although it has a similar structure to the other propanesulfonic acids and was selected as a highly water soluble buffering reagent with an optimum buffering pH of 10.4 and minimal reactivity with enzymes or proteins, minimal salt effects.

Solubility
Soluble in water (100 g/L) at 20°C, and methanol (1%).

Notes
Store at room temperature. Ensure good ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Concentration 0.5 M
pH 10.5
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material CAPS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.