missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 0.5M buffer soln., pH 11.0

Product Code. 15405409
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405409 250 mL 250mL
15415409 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405409 Supplier Thermo Scientific Chemicals Supplier No. J63138.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Concentration 0.5 M
pH 11.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material CAPS
compliance-icons
Product Identifier
  • CAPS, 0.5M buffer solution, pH 11.0
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-(cyclohexylamino)-1-propanesulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.