missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 99%, for biochemistry

Product Code. 10347830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347830 100 g Glass bottle 100g
10331034 250 g Glass bottle 250g
10390544 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10347830 Supplier Thermo Scientific Chemicals Supplier No. 172621000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym 3-Cyclohexylamino-1-propanesulfonic acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Absorbance (0.1 M in H2O) at 0nm
Absorbance Conditions (0.1M in H2O)
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 5% max.
Linear Formula C6H11NH(CH2)3SO3H
Packaging Glass bottle
Merck Index 15, 1769
Formula Weight 221.31
Percent Purity ≥98.5% (on dry substance)
Grade Biochemical
Chemical Name or Material CAPS, 99%
Show More Show Less
compliance-icons
Product Identifier
  • CAPS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.