Learn More
Erlab™ Captair by Erlab™ Stainless Steel Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Stainless Steel Work Surfaces are for use with Captair PCR work stations.
2141.20 € - 2952.10 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
13416583
|
Erlab™
I0116050001 |
CaptairFlow 391 ductless fume hoods |
2141.20 €
Each |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13436583
|
Erlab™
I0118050001 |
CaptairFlow 483 ductless fume hoods |
2480.40 €
Each |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13426583
|
Erlab™
I0120050001 |
CaptairFlow 714 ductless fume hoods |
2952.10 €
Each |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- High chemical and mechanical resistance
- Rounded corners to facilitate cleaning operations
- Built in spill tray
Specifications
Stainless Steel |