Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Erlab™ Captair Smart Trespa™ Work Surface is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
Brand:  Erlab™ I0161030001
Additional Details : Weight : 17.00000kg
 View more versions of this product
Product Code. 15575154
  • 1028.20 € / Each
Estimated Shipment: 27-08-2024
to see stock.
Description
Description
  • Made of phenolic resin with ergonomic arm for a comfortable working position
  • High chemical and mechanical resistance
  • Ideal for precision weighing operations
Specifications
Specifications
Smart 391 Fume Hood
510 mm
Work Surface
Spill Tray
900 mm
Phenolic Resin