missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZA2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92643-0.1ml
This item is not returnable.
View return policy
Description
CAPZA2 Polyclonal antibody specifically detects CAPZA2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CAPZA2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| CAPPA2CAPZ, capping protein (actin filament) muscle Z-line, alpha 2, CapZ alpha-2, F-actin capping protein alpha-2 subunit, F-actin-capping protein subunit alpha-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-286 of human CAPZA2 (NP_006127.1). MADLEEQLSDEEKVRIAAKFIIHAPPGEFNEVFNDVRLLLNNDNLLREGAAHAFAQYNLDQFTPVKIEGYEDQVLITEHGDLGNGKFLDPKNRICFKFDHLRKEATDPRPCEVENAVESWRTSVETALRAYVKEHYPNGVCTVYGKKIDGQQTIIACIESHQFQAKNFWNGRWRSEWKFTITPSTTQVVGILKIQVHYYEDGNVQLVSHKDIQDSLTVSNEVQTAKEFIKIVEAAENEYQTAISENYQTMSDTTFKALRRQLPVTRTKIDWNKILSYKIGKEMQNA | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 830 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction