missing translation for 'onlineSavingsMsg'
Learn More

Carbachol, 99%, Thermo Scientific™

Carbachol, CAS # 51-83-2, is a parasympathomimetic that acts on acetylcholine receptors.

39.20 € - 386.00 €

Chemical Identifiers

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-(trimethylazaniumyl)ethyl carbamate chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15438859
SDS View Documents
Thermo Scientific Alfa Aesar
L06674.03
1g
39.20 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15448859
SDS View Documents
Thermo Scientific Alfa Aesar
L06674.06
5g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15458859
SDS View Documents
Thermo Scientific Alfa Aesar
L06674.14
25g
386.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Carbachol is an ammonium salt and a choline ester that acts on both muscarinic and nicotinic receptors
• Carbachol activates the sympathetic nervous system; it leads to intracellular increases in Ca2+ and activates associated signaling pathways
• Carbachol is not degraded by the cholinesterases
Application
• Carbachol can inhibit apoptotic cell death, making it useful for the study of cell survival and growth in laboratory settings
• Carbachol can be used for laboratory research of any biological pathway that depends on acetylcholine, including sleep cycles and motor control
Specifications

Chemical Identifiers

51-83-2
182.65
AIXAANGOTKPUOY-UHFFFAOYSA-N
5831
2-(trimethylazaniumyl)ethyl carbamate chloride
C6H15ClN2O2
MFCD00012011
carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
CHEBI:3385
[Cl-].C[N+](C)(C)CCOC(N)=O

Specifications

51-83-2
MFCD00012011
3917459
14,1779
Soluble in water and dimethyl sulfoxide.
[Cl-].C[N+](C)(C)CCOC(N)=O
182.65
CHEBI:3385
99%
C6H15ClN2O2
UN2811
Hygroscopic
carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
AIXAANGOTKPUOY-UHFFFAOYSA-N
2-(trimethylazaniumyl)ethyl carbamate chloride
5831
182.65
Carbachol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H300-H500

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 200-127-3

missing translation for 'rtecsNumber' : GA0875000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Carbachol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2

Hazard Statement
  • H300-Fatal if swallowed.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

Supplemental information
  • MIXTURE LIST-Contain: Carbachol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.