missing translation for 'onlineSavingsMsg'
Learn More

Carbadox, 98+%

Product Code. 15473079
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473079 100 g 100g
15463079 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473079 Supplier Thermo Scientific Alfa Aesar Supplier No. J60284.22
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6804-07-5 | C13H10BrNO | 276.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

Solubility
Soluble in 1N NaOH (50 mg/ml), and water (partly).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6804-07-5
MDL Number MFCD00057293

Specifications

Melting Point 239°C to 240°C
Color Yellow
Flash Point 18°C (64°F)
Molecular Formula C11H10N4O4
Quantity 100 g
UN Number UN1325
Merck Index 14,1780
Synonym carbadox, unii-m2x04r2e2y, getroxel, mecadox, dsstox_cid_23913, dsstox_rid_80088, dsstox_gsid_43913, fortigro, karbadox czech, methyl n-e-1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene methyl iminocarbamate
Solubility Information Soluble in 1N NaOH (50mg/ml),and water (partly).
InChI Key BPMVRAQIQQEBLN-UHFFFAOYSA-N
SMILES COC(=O)N=NC=C1C=[N+](C2=CC=CC=C2N1O)[O-]
IUPAC Name methyl N-[(E)-(1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene)methyl]iminocarbamate
Molecular Weight (g/mol) 262.23
PubChem CID 5353472
Formula Weight 262.22
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Carbadox
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.