missing translation for 'onlineSavingsMsg'
Learn More

Carbamate Pesticides Mixture 638 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30323282
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323282 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323282 Supplier Dr. Ehrenstorfer Supplier No. DREA50000638ME
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Naphthol∣Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Methiocarb∣Methomyl∣Oxamyl∣Propoxur
CAS 90-15-3, 116-06-3, 1646-88-4, 1646-87-3, 63-25-2, 1563-66-2
Analyte or Component Names 1-Naphthol∣Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Methiocarb∣Methomyl∣Oxamyl∣Propoxur
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1-Naphthol 100 μg/mL∣Aldicarb 100 μg/mL∣Aldicarb-sulfone 100 μg/mL∣Aldicarb-sulfoxide 100 μg/mL∣Carbaryl 100 μg/mL∣Carbofuran 100 μg/mL∣Carbofuran-3-hydroxy 100 μg/mL∣Methiocarb 100 μg/mL∣Methomyl 100 μg/mL∣Oxamyl 100 μg/mL∣Propoxur 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.