missing translation for 'onlineSavingsMsg'
Learn More

Carbenicillin (Disodium Salt), Fisher BioReagents

Product Code. 10438923
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10438923 250 mg Amber Glass 250mg
10163193 1 g Amber Glass 1g
10396833 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10438923 Supplier Fisher BioReagents Supplier No. BP2648250
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy
Used for the selection of ampicillin-resistant transformed cells and for the study of penicillin-sensitive transpeptidases in cell wall biosynthesis. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4800-94-6
Molecular Formula C17H16N2Na2O6S
Molecular Weight (g/mol) 422.36
MDL Number MFCD00077683
InChI Key RTYJTGSCYUUYAL-YCAHSCEMSA-L
Synonym α-Carboxybenzylpenicillin
PubChem CID 20933
ChEBI CHEBI:34609
IUPAC Name disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Carbenicillin
Physical Form Solid
Melting Point 200°C
Packaging Amber Glass
Potency 770μg/mg min.
Water 6% Max.
Color White
Quantity 250 mg
Assay Percent Range ≥92 %
Boiling Point 738°C
Formula Weight 422.4
Percent Purity 92% min.
Chemical Name or Material Carbenicillin, Disodium Salt
Show More Show Less
compliance-icons
Product Identifier
  • Carbenicillin disodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.