missing translation for 'onlineSavingsMsg'
Learn More

Carbenicillin Disodium Salt, MP Biomedicals™

Product Code. 11460952 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460952 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460952 Supplier MP Biomedicals Supplier No. 02195092.2
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Displays antipseudomonal and antiproteal effects. Carbenicillin Disodium Salt, MP Biomedicals is a carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface

Carbenicillin is a semi-synthetic antibiotic that is an analog to ampicillin. It is derived from benzyl penicillin by substituting the a-carboxyl group in the benzyl side chain. It is completely ionized in the blood and is about 50% reversibly protein bound in plasma. Its tissue distribution is similar to other penicillins.

Susceptible bacteria include gram-positive and gram-negative bacteria abd pseudomonas.Though it has substantial in vitro activity against a variety of both gram-positive and gram-negative microorganisms, the most important aspect of its profile is in its antipseudomonal and antiproteal effect.

Carbenicillin Disodium Salt is used for selection of ampr transformed cells and to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.

Specifications

Physical Form Powder
CAS 4800-94-6
Potency >650μg/mg
Color White
Concentration Working Concentration: 105u/L (5-50μg/mL) in cell culture; approximately 200mg/kg in vivo.
Quantity 250 mg
Molecular Formula C17H16N2Na2O6S
MDL Number MFCD00077683
Synonym α-Carboxybenzylpenicillin
Solubility Information Soluble in water (50mg/mL) and alcohol
InChI Key RTYJTGSCYUUYAL-YCAHSCEMSA-L
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
IUPAC Name disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 422.36
PubChem CID 20933
ChEBI CHEBI:34609
Formula Weight 422.4
Grade Not Available
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Carbenicillin Disodium Salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Carbenicillin Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.