missing translation for 'onlineSavingsMsg'
Learn More

Carbidopa monohydrate, 99%

Product Code. 15698705 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15698705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15698705 Supplier Thermo Scientific Acros Supplier No. 461010010
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38821-49-7 | C10H14N2O4·H2O | 244.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38821-49-7
Loss on Drying 6.9% to 7.9% (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C10H14N2O4·H2O
Quantity 1 g
Synonym 3-3,4-dihydroxyphenyl-2-hydrazinyl-2-methylpropanoic acid hydrate, +-2-3,4-dihydroxybenzyl-2-hydrazinopropionic acid monohydrate, 3-3,4-bis oxidanyl phenyl-2-diazanyl-2-methyl-propanoic acid hydrate
InChI Key QTAOMKOIBXZKND-PPHPATTJSA-N
SMILES CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN.O
IUPAC Name (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate
Molecular Weight (g/mol) 244.25
PubChem CID 38101
ChEBI CHEBI:3395
Formula Weight 244.25
Percent Purity 99%
Chemical Name or Material Carbidopa monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Carbidopa monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Carbidopa

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.