missing translation for 'onlineSavingsMsg'
Learn More

Carbimazole, MedChemExpress

Product Code. 16492229
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492229 1 g 1g
16460998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492229 Supplier Medchem Express Supplier No. HYB0558
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Carbimazole is an imidazole antithyroid agent and can be used for the research of Graves' disease. Carbimazole plays its role due to its rapid conversion to methylmercapto imidazole (MMI) in vivo and can be converted to methimazole in vitro.

Carbimazole is an imidazole antithyroid agent and can be used for the research of Graves' disease. Carbimazole plays its role due to its rapid conversion to methylmercapto imidazole (MMI) in vivo and can be converted to methimazole in vitro.

Specifications

Chemical Name or Material Carbimazole
Physical Form Solid
CAS 22232-54-8
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C7H10N2O2S
Solubility Information DMSO : 100 mg/mL (536.97 mM; Need ultrasonic)
SMILES O=C(N1C=CN(C)C1=S)OCC
Molecular Weight (g/mol) 186.23
Formula Weight 186.23
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.