missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbofuran, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Carbofuran |
| CAS | 1563-66-2 |
| CAS Max % | ≤ 100% |
| Linear Formula | C12H15NO3 |
| Beilstein | 1428746 |
| Color | White |
| Melting Point | 153°C to 154°C |
| Synonym | 2,3-dihydro-2,2-dimethyl-7-benzofuranyl N-methylcarbamate |
| Molecular Weight (g/mol) | 221.3 g/mol |
| Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction