missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%, Solstice
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -112°C to -111°C (literature) |
| Absorbance | max. 0.50 at 388 nm, max. 1.0 at 386 nm, max. 0.01 at 750 nm, max. 0.01 500 nm, max. 0.05 at 410 nm, max. 0.10 at 403 nm, max. 0.25 at 394 nm |
| CAS Max % | 100.0000% |
| Density | 1.266 g/mL at 25 °C(lit.) |
| Quantity | 4 x 2 L |
| Flash Point | −30°C |
| Packaging | Glass Bottle |
| Linear Formula | CS2 |
| UN Number | UN1131 |
| Beilstein | 1098293 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?