missing translation for 'onlineSavingsMsg'
Learn More

Carbonate solution, 0.22M Carb./ 0.28M Bicarb. for Ion Chromatography, 100mg/L, H2O matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12982235 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12982235 250 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12982235 Supplier Fisher Chemical Supplier No. A 104 100
  • ONLINE EXCLUSIVE PRICE
    83.45 €
    64.55 € / 100mL
    Save 18.90 €
    22% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 497-19-8
Physical Form Liquid
Molecular Formula CNa2O3
Concentration 100 mg/L
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Type Single-component Standard Solution
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Solvent or Matrix Water
Molecular Weight (g/mol) 105.99
PubChem CID 10340
Quantity 250 mL
ChEBI CHEBI:29377
Chemical Name or Material Carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.