missing translation for 'onlineSavingsMsg'
Learn More

Carbonate solution, 0.5M Sodium Carbonate, for Ion Chromatography, H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12972235 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12972235 500 mL Bottle 500mL
12962235 100 mL Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12972235 Supplier Fisher Chemical Supplier No. A 103 500
  • ONLINE EXCLUSIVE PRICE
    150.00 €
    135.20 € / 500mL
    Save 14.80 €
    9% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Packaging Bottle
Solution Contains 0.5M Sodium Carbonate
Physical Form Liquid
Concentration 0.5 M
Type Single-component Standard Solution
IUPAC Name disodium carbonate
Solvent or Matrix Water
Quantity 500 mL
Chemical Name or Material Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.