missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase II/CA2 Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89522-100UL
This item is not returnable.
View return policy
Description
Carbonic Anhydrase II/CA2 Monoclonal specifically detects Carbonic Anhydrase II/CA2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Carbonic Anhydrase II/CA2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 1 | |
| Western Blot 1:10000-1:50000, Immunoprecipitation 1-4 ul/mg of lysate | |
| CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase | |
| Recombinant Human Carbonic Anhydrase II/CA2 protein | |
| 100 μg | |
| Hypoxia, Lipid and Metabolism | |
| 760 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction