Learn More
Carbonic Anhydrase VI Polyclonal Antibody
GREENER_CHOICE
Carbonic Anhydrase VI Polyclonal Antibody
Brand:  Invitrogen PA548022
Product Code. 15568512
  • 520.00 € / 100µg
Description
Carbonic Anhydrase VI Polyclonal Antibody for Western Blot, IP
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carbonic Anhydrase VI
Polyclonal
Unconjugated
CA6
CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase
Goat
Antigen Affinity Chromatography
RUO
765
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
P23280
CA6
Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase VI Gln18-Asn308.