missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase X Monoclonal Antibody (7H8F12E6), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA542996

 View more versions of this product

Product Code. 17719553

  • 449.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: Human CA10/CARPX

This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase X
Monoclonal
1 mg/mL
PBS with no preservative
Q9NS85
CA10
Recombinant Human CA10 protein.
100 μL
Primary
Human
Antibody
IgG1
ELISA
7H8F12E6
Unconjugated
CA10
2700029L05Rik; BB085816; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; HUCEP-15; UNQ533/PRO1076
Mouse
Protein A
RUO
56934
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.