Learn More
Carbonic Anhydrase XII/CA12 Antibody (CL0278) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52901PE
Product Code. 30294063
  • 737.00 € / 0.10mL
Description
Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carbonic Anhydrase XII/CA12
Monoclonal
PE
Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
This antibody was developed using a recombinant protein derived from O43570, with the exact immunogen sequence remaining proprietary.
0.1 mL
Lipid and Metabolism
771
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0278
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified