missing translation for 'onlineSavingsMsg'
Learn More

Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I), Rh 20%

Product Code. 11305968
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11305968

Brand: Thermo Scientific Alfa Aesar 010458.01

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydroformylation | CAS: 25470-96-6 | C24H22O3PRh | 492.32 g/mol

Hydroformylation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25470-96-6
Molecular Formula C24H22O3PRh
Molecular Weight (g/mol) 492.32
MDL Number MFCD00064611
InChI Key UGUXXRZSXZLSCY-MECAPONASA-M
IUPAC Name λ¹-rhodium(1+) methanidylidyneoxidanium triphenylphosphane (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].[C-]#[O+].C\C([O-])=C\C(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 0.25 g
Merck Index 14,356
Solubility Information Soluble in acetone and chlorinated solvents
Formula Weight 492.32
Concentration or Composition (by Analyte or Components) Rh 21%
Physical Form Crystals
Chemical Name or Material Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.