missing translation for 'onlineSavingsMsg'
Learn More

Carbonyl cyanide 3-chlorophenylhydrazone, 98%

Product Code. 11357056
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357056 500 mg 500mg
11367056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357056 Supplier Thermo Scientific Alfa Aesar Supplier No. L06932.ME
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 555-60-2 | C9H5ClN4 | 204.617 g/mol

Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.

Solubility
Insoluble in water. Soluble in DMSO (5 mg/ml), ethanol (1 mg/ml) and methanol (10 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-60-2
Molecular Formula C9H5ClN4
Molecular Weight (g/mol) 204.617
MDL Number MFCD00001848
InChI Key UGTJLJZQQFGTJD-UHFFFAOYSA-N
Synonym cccp, 3-chlorophenyl hydrazonomalononitrile, carbonylcyanide-3-chlorophenylhydrazone, carbonyl cyanide 3-chlorophenylhydrazone, carbonyl cyanide m-chlorophenyl hydrazone, carbonyl cyanide m-chlorophenylhydrazone, 3-chlorophenyl hydrazono malononitrile, m-chlorophenyl carbonylcyanide hydrazone, m-cl-ccp, carbonyl cyanide, 3-chlorophenyl hydrazone
PubChem CID 2603
ChEBI CHEBI:3259
IUPAC Name 2-[(3-chlorophenyl)hydrazinylidene]propanedinitrile
SMILES C1=CC(=CC(=C1)Cl)NN=C(C#N)C#N

Specifications

Melting Point ∽175°C (decomposition)
Odor Odorless
Quantity 500 mg
UN Number UN3439
Beilstein 1842102
Solubility Information Insoluble in water. Soluble in DMSO (5mg/ml),ethanol (1mg/ml) and methanol (10mg/ml).
Formula Weight 204.62
Percent Purity 98%
Chemical Name or Material Carbonyl cyanide 3-chlorophenylhydrazone
compliance-icons
Product Identifier
  • Carbonyl cyanide 3-chlorophenylhydrazone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Carbonyl cyanide 3-chlorophenylhydrazone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.