missing translation for 'onlineSavingsMsg'
Learn More

Carbonylchlorobis(triphenylphosphine)iridium(I)

Product Code. 11338198
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338198 0.25 g 0.25g
11348198 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338198 Supplier Thermo Scientific Alfa Aesar Supplier No. 039413.01
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14871-41-1 | C37H30ClIrOP2 | 780.26 g/mol

Carbonylchlorobis(triphenylphosphine)iridium(I) is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14871-41-1
Molecular Formula C37H30ClIrOP2
Molecular Weight (g/mol) 780.26
MDL Number MFCD00009590
InChI Key PEPSDKMVHYXIMJ-UHFFFAOYSA-M
Synonym carbonylchlorobis triphenylphosphine iridium i
PubChem CID 78076234
IUPAC Name iridium;methanone;triphenylphosphane;chloride
SMILES Cl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 215°C (decomposition)
Quantity 0.25 g
UN Number UN2811
Solubility Information Soluble in chloroform, toluene; Slightly soluble in acetone, alcohol
Formula Weight 780.27
Physical Form Crystalline
Chemical Name or Material Carbonylchlorobis(triphenylphosphine)iridium(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.