missing translation for 'onlineSavingsMsg'
Learn More

Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min

Product Code. 11316717
Click to view available options
Quantity:
0.1 g
1 g
5 g
Unit Size:
0.1g
1g
5g
This item is not returnable. View return policy

Product Code. 11316717

Brand: Thermo Scientific Alfa Aesar 010016.77

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation | CAS: 17185-29-4 | C55H46OP3Rh | 918.80 g/mol

Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17185-29-4
Molecular Formula C55H46OP3Rh
Molecular Weight (g/mol) 918.80
MDL Number MFCD00151644
InChI Key AACIZACVKFEETJ-UHFFFAOYSA-N
Synonym tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i
PubChem CID 132451836
IUPAC Name carbon monoxide;rhodium;triphenylphosphane
SMILES [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 138°C (decomposition)
Color Yellow
Density 1.33 g/mL
Odor Odorless
Linear Formula RhH(CO)[P(C6H5)3]3
Quantity 0.1 g
Sensitivity Moisture sensitive
Merck Index 14,9758
Solubility Information Soluble in hydrocarbons (e.g. benzene and toluene) with dissociation of phosphine ligands
Formula Weight 918.8
Concentration or Composition (by Analyte or Components) Rh 10.0% min.
Physical Form Powder
Chemical Name or Material Carbonylhydridotris(triphenylphosphine)rhodium(I)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.