missing translation for 'onlineSavingsMsg'
Learn More

Carboxyformamido Metazachlor Benzoic Acid, TRC

Product Code. 30180271
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180271 10 mg 10mg
30180182 50 mg 50mg
30179901 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180271 Supplier Toronto Research Chemicals Supplier No. TRCM85459010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1367578-41-3
Molecular Formula C14 H13 N3 O5
Molecular Weight (g/mol) 303.27
Synonym Metazachlor Metabolite BH 479-12, Benzoic acid, 2-[(carboxycarbonyl)(1H-pyrazol-1-ylmethyl)amino]-3-methyl-
IUPAC Name 3-methyl-2-[oxalo(pyrazol-1-ylmethyl)amino]benzoic acid
SMILES Cc1cccc(C(=O)O)c1N(Cn2cccn2)C(=O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H13N3O5/c1-9-4-2-5-10(13(19)20)11(9)17(12(18)14(21)22)8-16-7-3-6-15-16/h2-7H,8H2,1H3,(H,19,20)(H,21,22)
Formula Weight 303.0855
Chemical Name or Material Metazachlor Metabolite BH 479-12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.