Learn More
Carboxypeptidase A1/CPA1 Antibody (CL6629) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76509AF750
Product Code. 30287933
  • 737.00 € / 0.10mL
Description
Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carboxypeptidase A1/CPA1
Monoclonal
Alexa Fluor 750
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1
This antibody was developed using a recombinant protein derived from P15085, with the exact immunogen sequence remaining proprietary.
0.1 mL
Mast Cell Markers
1357
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL6629
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified