Learn More
Carboxypeptidase A1/CPA1 Antibody (CPA1/2712) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-79880
Product Code. 18294037
  • 599.00 € / 100µg
Description
Carboxypeptidase A1/CPA1 Monoclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carboxypeptidase A1/CPA1
Monoclonal
1 mg/mL
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1
Mouse
Protein A or G purified
RUO
Primary
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG2c κ
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
CPA1/2712
Unconjugated
10 mM PBS with No Preservative
CPA1
Recombinant full-length human Carboxypeptidase A1/CPA1 protein (Uniprot: P15085)
100 μg
Mast Cell Markers
Human
Purified