missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (rCPA1/8688), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20326-20ug
This item is not returnable.
View return policy
Description
Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Carboxypeptidase A1/CPA1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
| Recombinant full-length human Carboxypeptidase A1/CPA1 protein | |
| 20 μg | |
| Mast Cell Markers | |
| 1357 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rCPA1/8688 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction