missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA569156
Additional Details : Weight : 0.01000kg
Description
This target displays homology in the following species: Cow: 86%; Dog: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 93%; Rat: 93%; Yeast: 90%
Three different forms of human pancreatic procarboxypeptidase A have been isolated. This gene encodes a form which is obtained as a binary complex of a procarboxypeptidase A with proproteinase E and functions as a secretory granule metalloexopeptidase.Specifications
Carboxypeptidase A3 | |
Polyclonal | |
Unconjugated | |
CPA3 | |
Carboxypeptidase A3; carboxypeptidase A3 (mast cell); carboxypeptidase A3, mast cell; Cpa3; Mast cell carboxypeptidase A; MC-CPA; mMC-CPA; R-CPA; RMC-CP; tissue carboxypeptidase A | |
Rabbit | |
Affinity chromatography | |
RUO | |
12873 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P15089 | |
CPA3 | |
synthetic peptide corresponding to a region of mouse Carboxypeptidase A3 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |