Learn More
Cardiac Troponin T Antibody (OTI8B5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44702
Product Code. 30306602
  • 735.00 € / 100µg
Description
Cardiac Troponin T Monoclonal antibody specifically detects Cardiac Troponin T in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cardiac Troponin T
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
7139
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry, Western Blot
OTI8B5
Western Blot 1:2000, Immunohistochemistry 1:150
Cardiac Muscle Troponin T, Cardiomyopathy, Dilated 1D (Autosomal Dominant), Cardiomyopathy, Hypertrophic, CMD1D, CMH2, CMPD2, cTnT, LVNC6, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle, Troponin T2, Cardiac Type, Truncated Cardiac Troponin T
Native troponin complex
100 μg
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized