missing translation for 'onlineSavingsMsg'
Learn More

Carminic acid (E120) 100 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer

Product Code. 30320275
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320275 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320275 Supplier Dr. Ehrenstorfer Supplier No. DREA11044000MC100
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carminic Acid
CAS 1260-17-9
Analyte or Component Names Carminic Acid
Shipping Condition Room Temperature
Molecular Formula C22 H20 O13
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2-Anthracenecarboxylic acid, 7-β-D-glucopyranosyl-9,10-dihydro-3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-, 2-Anthroic acid, 7-D-glucopyranosyl-9,10-dihydro-3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo- (7CI), Carminic acid (6CI,8CI), 7-β-D-Glucopyranosyl-9,10-dihydro-3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-2-anthracenecarboxylic acid, C.I. 75470, C.I. Natural Red 4, Carmine 5297, Cochineal Red PWD, E 120, E 120 (dye), NSC 326224, Natural Red 2180, Natural Red 4, San Red 1, San-Ei Gen San Red 1, Sun Red 1, Sun Red No. 1
InChI Formula InChI=1S/C22H20O13/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34)/t7-,14-,19+,20-,21+/m1/s1
SMILES Cc1c(C(=O)O)c(O)cc2C(=O)c3c(O)c(O)c([C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c(O)c3C(=O)c12
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid
Solvent or Matrix Acetonitrile/Methanol
Molecular Weight (g/mol) 492.39
Quantity 1 mL
Formula Weight 492.0904 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.