missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cas9 Monoclonal Antibody (OTI1C8), TrueMAB™, OriGene
Beskrivelse
CRISPR systems are adaptable immune mechanisms which are present in many bacteria to protect themselves from foreign nucleic acids, such as viruses, transposable elements, or plasmids. Recently, the CRISPR/Cas9 system from S. pyogenes has been adapted for inducing sequence-specific double stranded breaks and targeted genome editing. This system is unique and flexible due to its dependence on RNA as the element that targets the nuclease to a desired DNA sequence. It can be used to induce indel mutations, specific sequence replacements/insertions, and large deletions or genomic rearrangements at any desired location in the genome. In addition, Cas9 can also be used to mediate upregulation of specific endogenous genes or to alter histone modifications or DNA methylation.
Tekniske data
Tekniske data
| Antigen | CAS9 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | OTI1C8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene Alias | Cas9Cas9; Cas9 |
| Host Species | Mouse |
| Immunogen | Human recombit protein fragment corresponding to amino acids 1-1166 of human CAS9 produced in E.coli. |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?