missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-14 Antibody (001), Novus Biologicals™
Rabbit Monoclonal Antibody
275.00 € - 453.00 €
Specifications
| Antigen | Caspase-14 |
|---|---|
| Clone | 001 |
| Dilution | Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668368
|
Novus Biologicals
NBP3-05780-100ul |
100 μg |
453.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638388
|
Novus Biologicals
NBP3-05780-50ul |
50 μg |
275.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-14 Monoclonal antibody specifically detects Caspase-14 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Caspase-14 | |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Caspase-14 (Accession#: NP_036246.1; Ser2-Gln242). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 001 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Caspases | |
| 0.2 um filtered solution in PBS | |
| 23581 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title