missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Antibody - (active/cleaved), Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 48 publications
232.00 € - 484.00 €
Specifications
| Antigen | Caspase-3 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Flow Cytometry reported in scientific literature (PMID 28287161), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29963272), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen reported in scientific literature (PMID 29864441), Immunoblotting reported in scientific literature (PMID 27912032), Flow (Intracellular) reported in scientific literature (PMID 24804954) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18745403
|
Novus Biologicals
NB100-56113SS |
0.025 mL |
232.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749613
|
Novus Biologicals
NB100-56113 |
0.05 mL |
484.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular).Specifications
| Caspase-3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Caspases, Phospho Specific | |
| Neat whole antisera with 0.05% Sodium Azide | |
| 836 | |
| This Caspase-3 Antibody - (active/cleaved) was developed against catalytically active human caspase-3 protein. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 31.7 kDa |
| Western Blot 1:1000-1:2000, Flow Cytometry reported in scientific literature (PMID 28287161), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29963272), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen reported in scientific literature (PMID 29864441), Immunoblotting reported in scientific literature (PMID 27912032), Flow (Intracellular) reported in scientific literature (PMID 24804954) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Gerbil | |
| Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| CASP3 | |
| IgG | |
| Unpurified | |
| Preferentially detects active caspase-3 (large subunit: ∼14-21 kDa, and small subunit: ∼10 kDa). However, it may also detect pro-caspase-3 (∼32 kDa) in some cell or tissue systems. Nuclear immunostaining of caspase-3 is considered be an indication of active/cleaved caspase-3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title