missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Antibody (CPP32 4-1-18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
226.00 € - 494.00 €
Specifications
| Antigen | Caspase-3 |
|---|---|
| Clone | CPP32 4-1-18 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Simple Western 1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunoprecipitation 2 ug/ mg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 31242448) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18709383
|
Novus Biologicals
NB500-210SS |
0.05 mL |
226.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740204
|
Novus Biologicals
NB500-210 |
0.2 mL |
494.00 €
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Caspase-3 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Caspases | |
| PBS with 0.02% Sodium Azide | |
| Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| CASP3 | |
| IgG2a κ | |
| Protein G purified | |
| 31.7 kDa |
| CPP32 4-1-18 | |
| Western Blot 1:500-1:1000, Simple Western 1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunoprecipitation 2 ug/ mg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 31242448) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat | |
| P42574 | |
| 836 | |
| This Caspase-3 Antibody (CPP32 4-1-18) was developed against full-length recombinant human Caspase 3 [UniProt# P42574]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title