missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00 € - 470.00 €
Specifications
| Antigen | Caspase 5 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665750
|
Novus Biologicals
NBP2-92338-0.02ml |
0.02 mL |
226.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603511
|
Novus Biologicals
NBP2-92338-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase 5 Polyclonal antibody specifically detects Caspase 5 in Human samples. It is validated for Western BlotSpecifications
| Caspase 5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Caspases | |
| PBS with 50% glycerol, pH7.3. | |
| 838 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CASP-5, caspase 5, apoptosis-related cysteine peptidase, caspase 5, apoptosis-related cysteine protease, caspase-5, EC 3.4.22, EC 3.4.22.58, ICE(rel)III, ICE(rel)-III, ICEREL-III, ICH3, ICH-3, MGC141966, Protease ICH-3, Protease TY, TY protease | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-250 of human Caspase-5 (NP_001129584). QMFTQTLLNMDQKITSVKPLLQIEAGPPESAESTNILKLCPREEFLRLCKKNHDEIYPIKKREDRRRLALIICNTKFDHLPARNGAHYDIVGMKRLLQGLGYTVVDEKNLT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title