missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Caspase 6 Recombinant Rabbit Monoclonal Antibody (K01_1K32)

Product Code. 30830458
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830458 50 μL
30824622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830458 Supplier Invitrogen™ Supplier No. MA564727
 Restricted Item
Estimated Shipment: 09-10-2026
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: AASVYTLPAG ADFLMCYSVA EGYYSHRETV NGSWYIQDLC EMLGKYGSSL EFTELLTLVN RKVSQRRVDF CKDPSAIGKK QVPCFASMLT KKLHFFPKSN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%, Pig - 98%, Cynomolgus monkey - 100%.

Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
TRUSTED_SUSTAINABILITY

Spécification

Antigen Caspase 6
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1K32
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Casp6
Gene Accession No. O35397, P55212
Gene Alias apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2
Gene Symbols Casp6
Host Species Rabbit
Immunogen Recombinant protein of human Caspase-6 (aa 194-293).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83584, 839
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.