missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody - BSA Free, Novus Biologicals™
CD4 (cluster of differentiation 4), also known as L3T4 or T4, is a 55 kDa single chain type I transmembrane glycoprotein belonging to the immunoglobin (Ig) superfamily.
309.00 € - 635.00 €
Specifications
| Antigen | CD4 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 ug/ml, Simple Western 1:100, Flow Cytometry 5 ug/ml, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18786243
|
Novus Biologicals
NBP1-19371SS |
0.025 mL |
309.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769863
|
Novus Biologicals
NBP1-19371 |
0.1 mL |
635.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Polyclonal specifically detects CD4 in Human, Mouse, Rat, Porcine, Canine, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC.Specifications
| CD4 | |
| Western Blot 1 ug/ml, Simple Western 1:100, Flow Cytometry 5 ug/ml, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| CD4 | |
| IgG | |
| Affinity Purified | |
| Rabbit reactivity reported in the scientific literature (PMID: 2212730). Customer feedback positive on canine. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Canine, Rabbit | |
| P01730, P06332 | |
| 920 | |
| A synthetic peptide made to an C-terminal region of the human CD4 Antibody protein (within residues 400-458). [Swiss-Prot P01730] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title