missing translation for 'onlineSavingsMsg'
Learn More

Caspofungin diacetate, MedChemExpress

Product Code. 16487258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
25mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487258

Brand: Medchem Express HY17006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Caspofungin (MK-0991) diacetate is a potent antifungal agent. Caspofungin diacetate inhibits the synthesis of the fungal cell wall component β-(l,3)-D-glucan.

Caspofungin (MK-0991) diacetate is a potent antifungal agent. Caspofungin diacetate inhibits the synthesis of the fungal cell wall component β-(l,3)-D-glucan.

Specifications

Chemical Name or Material Caspofungin diacetate
Physical Form Solid
CAS 179463-17-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C56H96N10O19
For Use With (Application) COVID-19-immunoregulation
Synonym MK-0991 diacetateL-743872 diacetate
Solubility Information H2O : ≥ 100 mg/mL (82.41 mM) ∣DMSO : 100 mg/mL (82.41 mM; Need ultrasonic)
SMILES O=C([C@](NC([C@](NC([C@@](C[C@H]1O)([H])N(C1)C2=O)=O)([H])[C@H](O)[C@H](C(C=C3)=CC=C3O)O)=O)([H])[C@H](O)CCN)N(CC[C@@H]4O)[C@]4([H])C(N[C@@H]([C@@H](C[C@@H](C(N[C@@]2([H])[C@H](O)C)=O)NC(CCCCCCCC[C@@H](C)C[C@@H](C)CC)=O)O)NCCN)=O.OC(C)=O.OC(C)=O
Molecular Weight (g/mol) 1213.42
Formula Weight 1213.42
Percent Purity 95.9%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.