missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspr1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-91994-0.1ml
This item is not returnable.
View return policy
Description
Caspr1 Polyclonal antibody specifically detects Caspr1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Caspr1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Caspr, Caspr1, CASPRcaspr1, CNTNAP, contactin associated protein 1, neurexin 4, Neurexin IV, Neurexin-4, NRXN4contactin-associated protein 1, P190 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human Caspr1 (NP_003623.1). WGYYGCDEELVGPLYARSLGASSYYSLLTAPRFARLHGISGWSPRIGDPNPWLQIDLMKKHRIRAVATQGSFNSWDWVTRYMLLYGDRVDSWTPFYQRGHNSTFFGNVNESAVVRHDLHFHFTARYIRIVPLAWNPRGKIGLRLGLYGCPYKADILYFDGDDAISYRFPRGVSRSLWDVFAFSFKTEEKDGLLLHAEGAQGDYVTLELEGAHLLLHMSLGSSPIQPRPGHTTVSAGGVLNDQHWHYVRVDRFGRDVNFTLDGYVQRFILNGDFERLNLDTE | |
| 0.1 mL | |
| Neuroscience | |
| 8506 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction