missing translation for 'onlineSavingsMsg'
Learn More

Catechol, 99+%

Product Code. 10542354 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Fibre drum
Glass bottle
Plastic Bottle
Unit Size:
100g
10kg
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10542354

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-80-9 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-80-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002188
InChI Key YCIMNLLNPGFGHC-UHFFFAOYSA-N
Synonym pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
PubChem CID 289
ChEBI CHEBI:18135
IUPAC Name benzene-1,2-diol
SMILES OC1=CC=CC=C1O

Specifications

Melting Point 103.0°C to 106.0°C
Color Brown-Gray to White
Boiling Point 245.0°C
Flash Point 131°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic Bottle
Quantity 2.5 kg
Merck Index 15,8111
Solubility Information Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol, benzene, chloroform, ether, and pyridine and aqueous solutions
Viscosity 2.38 mm2/s (110°C)
Formula Weight 110.11
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material Catechol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Catechol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.