missing translation for 'onlineSavingsMsg'
Learn More

Catechol-O,O-diacetic acid, 97%

Product Code. 11391417
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391417 50 g 50g
11381417 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391417 Supplier Thermo Scientific Alfa Aesar Supplier No. L01843.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5411-14-3 | C10H10O6 | 226.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5411-14-3
Molecular Formula C10H10O6
Molecular Weight (g/mol) 226.184
MDL Number MFCD00014353
InChI Key PPZYHOQWRAUWAY-UHFFFAOYSA-N
Synonym 1,2-phenylenedioxydiacetic acid, o-phenylenedioxydiacetic acid, 2,2'-1,2-phenylenebis oxy diacetic acid, catechol-o,o-diacetic acid, 2-2-carboxymethoxy phenoxy acetic acid, 2,2'-1,2-phenylenebis oxy bisacetic acid, 2-carboxymethoxy phenoxyacetic acid, 2,2'-benzene-1,2-diylbis oxy diacetic acid, acmc-1alcz, catechol-o,o'-diacetic acid
PubChem CID 79427
IUPAC Name 2-[2-(carboxymethoxy)phenoxy]acetic acid
SMILES C1=CC=C(C(=C1)OCC(=O)O)OCC(=O)O

Specifications

Melting Point 177°C to 181°C
Quantity 50 g
Beilstein 2057228
Formula Weight 226.18
Percent Purity 97%
Chemical Name or Material Catechol-O,O-diacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.