missing translation for 'onlineSavingsMsg'
Learn More

Catechol, TRC

Product Code. 30134725
Change view
Click to view available options
Quantity:
1 g
10 g
250 g
Unit Size:
10g
1g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134725 1 g 1g
30041297 10 g 10g
30006903 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134725 Supplier Toronto Research Chemicals Supplier No. TRCC2151751G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-80-9
Molecular Formula C6 H6 O2
Molecular Weight (g/mol) 110.11
Synonym Pyrocatechol (8CI), 1,2-Dihydroxybenzene, 1,2-Hydroxybenzene, 2-Hydroxyphenol, C.I. 76500, C.I. Oxidation Base 26, Catechol, Catechol (phenol), Durafur Developer C, Fouramine PCH, Fourrine 68, NSC 1573, Oxyphenic acid, Pelagol Grey C, Phthalhydroquinone, Phthalic alcohol, Pyrocatechin, Pyrocatechine, Z 1, Z 1 (inhibitor), o-Benzenediol, o-Dihydroxybenzene, o-Dioxybenzene, o-Hydroquinone, o-Hydroxyphenol, o-Phenylenediol
IUPAC Name benzene-1,2-diol
SMILES Oc1ccccc1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H
Formula Weight 110.0368
Chemical Name or Material Catechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.