missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin, beta (CTNNB1) Mouse, Clone: CTNNB1/2098, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1499-MSM11-P1
This item is not returnable.
View return policy
Specifications
| Catenin, beta (CTNNB1) | |
| Cell surface and cytoplasmic | |
| CTNNB1/2098 | |
| Unconjugated | |
| Mouse | |
| 92kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 1499 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human beta-Catenin (CTNNB1) protein | |
| HeLa or MCF-7 cells. Breast carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction