missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB400-152
This item is not returnable.
View return policy
Description
Cathepsin E Polyclonal specifically detects Cathepsin E in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cathepsin E | |
| Polyclonal | |
| Western Blot 1:2000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400 | |
| CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
| 1510 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| CTSE | |
| A synthetic peptide made to the C-terminus of mouse Cathepsin E [UniProt# P70269] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction