missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98303-100ul
This item is not returnable.
View return policy
Description
Cathepsin E Polyclonal antibody specifically detects Cathepsin E in Mouse samples. It is validated for ELISA
Specifications
| Cathepsin E | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
| Produced in rabbits immunized with purified, recombinant Mouse Cathepsin E (Accession#: NP_031825.2; Met 1-Pro 397) | |
| 100 μg | |
| Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
| 1510 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction