missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cathepsin S Monoclonal Antibody (248718)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA560935

Product Code. 30534239

  • 589.00 € / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cathepsin S
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P25774
CTSS
Mouse myeloma cell line NS0-derived recombinant human Cathepsin S, Gln17-Ile331 (predicted).
500 μg
Primary
Human
Antibody
IgG2b
Immunoprecipitation, Western Blot
248718
Unconjugated
CTSS
Cat S; cathepsin S; Cats; CTSS
Rat
Protein A/G
RUO
1520
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.