missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR1 Mouse anti-Human, Clone: 2D6, Novus Biologicals™
Mouse Monoclonal Antibody
294.00 € - 486.00 €
Specifications
| Antigen | CBR1 |
|---|---|
| Clone | 2D6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18264533
|
Novus Biologicals
NBP2-59397-50UL |
50 μL |
294.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18242925
|
Novus Biologicals
NBP2-59397-100UL |
100 μL |
486.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBR1 Monoclonal specifically detects CBR1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| CBR1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 873 | |
| Recombinant human CBR1 (1-277aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2D6 | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 15-hydroxyprostaglandin dehydrogenase [NADP+], carbonyl reductase (NADPH) 1, carbonyl reductase (NADPH) 1, EC 1.1.1.18410EC 1.1.1.197,15-hydroxyprostaglandin dehydrogenase, carbonyl reductase [NADPH] 1, carbonyl reductase 1, CBR, CRN, EC 1.1.1.184, EC 1.1.1.189, hCBR1, NADPH-dependent carbonyl reductase 1, Prostaglandin 9-ketoreductase, Prostaglandin-E(2) 9-reductase, SDR21C1, short chain dehydrogenase/reductase family 21C, member 1 | |
| CBR1 | |
| IgG2a κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title